ENA371464727 Enamine

[6-(2,2,2-trifluoroethoxy)pyridin-2-yl]methanamine

  • MDL number MFCD16704204
  • Cas Number : 1250054-65-9
  • Molecular Weight : 206.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [6-(2,2,2-trifluoroethoxy)-2-pyridinyl]methanamine
Inchi Code 1S/C8H9F3N2O/c9-8(10,11)5-14-7-3-1-2-6(4-12)13-7/h1-3H,4-5,12H2
Inchi Key FEKKEZOYDREICU-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-90365

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P310,P312,P330,P332+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-90365
pictograms GHS05,GHS07
hstatements H302,H315,H318,H335
Availability details for
Quantity
  • Availability
  • Add to Cart