ENA371464731 Enamine

2-[(1-ethyl-1H-1,3-benzodiazol-2-yl)sulfanyl]acetohydrazide

  • MDL number MFCD01914720
  • Cas Number : 306953-52-6
  • Molecular Weight : 250.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-110732

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110732
Availability details for
Quantity
  • Availability
  • Add to Cart