ENA371464733 Enamine

2-[(1-benzyl-1H-1,3-benzodiazol-2-yl)sulfanyl]acetohydrazide

  • MDL number MFCD01914539
  • Cas Number : 306951-60-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110733

Safety Information

msds https://www.enaminestore.com/catalog/EN300-110733
Availability details for
Quantity
  • Availability
  • Add to Cart