ENA371464738 Enamine

2-({1-[(2-chlorophenyl)methyl]-1H-1,3-benzodiazol-2-yl}sulfanyl)acetohydrazide

  • MDL number MFCD07364282
  • Cas Number : 893726-07-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110737

Safety Information

msds https://www.enaminestore.com/catalog/EN300-110737
Availability details for
Quantity
  • Availability
  • Add to Cart