ENA371464744 Enamine

4-methyl-1,2-oxazol-5-amine

  • MDL number MFCD11878418
  • Cas Number : 35143-75-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 59...61
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96607

Safety Information

msds https://www.enaminestore.com/catalog/EN300-96607
Availability details for
Quantity
  • Availability
  • Add to Cart