ENA371464754 Enamine

1-(3,3-diethoxypropoxy)-2-fluorobenzene

  • MDL number MFCD18598384
  • Cas Number : 1394040-87-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98957

Safety Information

msds https://www.enaminestore.com/catalog/EN300-98957
Availability details for
Quantity
  • Availability
  • Add to Cart