ENA372572362 Enamine

2,3,4,5,6-pentafluorophenyl 5-bromo-4-methyl-1,3-thiazole-2-sulfonate

  • MDL number MFCD22391915
  • Cas Number : 1421605-11-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104043

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-104043
Availability details for
Quantity
  • Availability
  • Add to Cart