ENA372572367 Enamine

5-bromo-4-chlorothiophene-2-sulfonyl chloride

  • MDL number MFCD00052093
  • Cas Number : 1307307-49-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104411

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-104411
Availability details for
Quantity
  • Availability
  • Add to Cart