ENA372572428 Enamine

2-chloro-N-[5-methyl-2-(5-methyl-4H-1,2,4-triazol-3-yl)phenyl]acetamide

  • MDL number MFCD22391984
  • Cas Number : 1421601-84-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-107840

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107840
Availability details for
Quantity
  • Availability
  • Add to Cart