ENA372572429 Enamine

1,2,5-thiadiazol-3-amine hydrochloride

  • MDL number MFCD22421767
  • Cas Number : 6595-67-1
  • Molecular Weight : 137.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1,2,5-thiadiazol-3-amine hydrochloride
Inchi Code 1S/C2H3N3S.ClH/c3-2-1-4-6-5-2;/h1H,(H2,3,5);1H
Inchi Key MQVGZOHLRZVLKO-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-107860

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-107860
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart