ENA372572492 Enamine

3-(3,3,3-trifluoropropoxy)prop-1-ene

  • MDL number MFCD22392061
  • Cas Number : 1421602-07-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110047

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110047
Availability details for
Quantity
  • Availability
  • Add to Cart