ENA372572494 Enamine

5-(1-methyl-1H-imidazol-2-yl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD22392067
  • Cas Number : 1421601-89-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110366

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110366
Availability details for
Quantity
  • Availability
  • Add to Cart