ENA372572517 Enamine

potassium (2S)-1-(methylcarbamoyl)pyrrolidine-2-carboxylate

  • MDL number MFCD22392097
  • Cas Number : 1423040-84-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111430

Safety Information

msds https://www.enaminestore.com/catalog/EN300-111430
Availability details for
Quantity
  • Availability
  • Add to Cart