ENA372572519 Enamine

3-amino-1-cyclopentyl-1H-pyrazole-4-carbothioamide

  • MDL number MFCD22392102
  • Cas Number : 1421602-06-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111505

Safety Information

msds https://www.enaminestore.com/catalog/EN300-111505
Availability details for
Quantity
  • Availability
  • Add to Cart