ENA372572530 Enamine

methyl 5-(1-chloroethyl)-3-methylfuran-2-carboxylate

  • MDL number MFCD22392126
  • Cas Number : 1421601-77-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111665

Safety Information

msds https://www.enaminestore.com/catalog/EN300-111665
Availability details for
Quantity
  • Availability
  • Add to Cart