ENA372572532 Enamine

1-methyl-1-[3-(methylsulfanyl)phenyl]thiourea

  • MDL number MFCD22392130
  • Cas Number : 1421606-02-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111707

Safety Information

msds https://www.enaminestore.com/catalog/EN300-111707
Availability details for
Quantity
  • Availability
  • Add to Cart