ENA372572534 Enamine

5-(4-chloro-3-ethylphenoxy)-N-oxo-1,4-dihydropyrazine-2-carboximidamide

  • MDL number MFCD22392136
  • Cas Number : 1421604-84-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-111854

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111854
Availability details for
Quantity
  • Availability
  • Add to Cart