ENA372572538 Enamine

1-N,1-N,3-N-trimethylbenzene-1,3-diamine dihydrochloride

  • MDL number MFCD22392144
  • Cas Number : 84995-22-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111882

Safety Information

msds https://www.enaminestore.com/catalog/EN300-111882
Availability details for
Quantity
  • Availability
  • Add to Cart