ENA372572539 Enamine

5-(pyridin-3-yl)thiophene-2-carboxylic acid hydrochloride

  • MDL number MFCD22392145
  • Cas Number : 1421602-69-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111928

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111928
Availability details for
Quantity
  • Availability
  • Add to Cart