ENA372572540 Enamine

(butan-2-yl)(methoxy)amine hydrochloride

  • MDL number MFCD22392146
  • Cas Number : 1421602-73-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111944

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111944
Availability details for
Quantity
  • Availability
  • Add to Cart