ENA372572548 Enamine

3-amino-5-(4-aminophenyl)-5-methylimidazolidine-2,4-dione

  • MDL number MFCD22392159
  • Cas Number : 1421604-88-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112049

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112049
Availability details for
Quantity
  • Availability
  • Add to Cart