ENA372572549 Enamine

1-(3-chloropropyl)-2-methyl-1H-imidazole hydrochloride

  • MDL number MFCD22392161
  • Cas Number : 1421601-58-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112096

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112096
Availability details for
Quantity
  • Availability
  • Add to Cart