ENA372572552 Enamine

2-(2-cyclopentylethyl)pyrrolidine hydrochloride

  • MDL number MFCD22392166
  • Cas Number : 1421602-09-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112160

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112160
Availability details for
Quantity
  • Availability
  • Add to Cart