ENA372572558 Enamine

4-(carbamimidoylmethoxy)-N-cyclopentylbenzamide hydrochloride

  • MDL number MFCD22392176
  • Cas Number : 1421604-21-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112230

Safety Information

msds https://www.enaminestore.com/catalog/EN300-112230
Availability details for
Quantity
  • Availability
  • Add to Cart