ENA372572559 Enamine

ethyl 4-(pyridin-3-yl)-1,3-thiazole-2-carboxylate hydrobromide

  • MDL number MFCD22392177
  • Cas Number : 1421602-11-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112231

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112231
Availability details for
Quantity
  • Availability
  • Add to Cart