ENA372572563 Enamine

4-(methylamino)benzene-1-carboximidamide dihydrochloride

  • MDL number MFCD22392182
  • Cas Number : 1421604-83-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112292

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112292
Availability details for
Quantity
  • Availability
  • Add to Cart