ENA372572571 Enamine

2-(4-nitro-1H-pyrazol-1-yl)ethan-1-amine hydrochloride

  • MDL number MFCD22392196
  • Cas Number : 1421602-82-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112448

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112448
Availability details for
Quantity
  • Availability
  • Add to Cart