ENA372572576 Enamine

3-(benzyloxy)-2,6-difluorobenzoic acid

  • MDL number MFCD22392201
  • Cas Number : 1421604-28-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112474

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112474
Availability details for
Quantity
  • Availability
  • Add to Cart