ENA372572577 Enamine

methyl 3-(1-hydroxy-2-methoxy-2-oxoethyl)benzoate

  • MDL number MFCD22392202
  • Cas Number : 947590-66-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112505

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112505
Availability details for
Quantity
  • Availability
  • Add to Cart