ENA372572586 Enamine

2,2-difluoro-2-(pyridin-2-yl)acetic acid hydrochloride

  • MDL number MFCD22392210
  • Cas Number : 1421603-45-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112605

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112605
Availability details for
Quantity
  • Availability
  • Add to Cart