ENA372572590 Enamine

benzyl 3-aminobicyclo[2.2.1]heptane-2-carboxylate hydrochloride

  • MDL number MFCD22392214
  • Cas Number : 1421604-37-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112666

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112666
Availability details for
Quantity
  • Availability
  • Add to Cart