ENA372572591 Enamine

9-ethyl-1,4-dioxa-7-azaspiro[4.4]nonane

  • MDL number MFCD22392215
  • Cas Number : 1421603-46-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112668

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112668
Availability details for
Quantity
  • Availability
  • Add to Cart