ENA372572595 Enamine

N-[(5-ethyl-1,3-oxazol-2-yl)methyl]-N-(pyrrolidin-3-yl)acetamide hydrochloride

  • MDL number MFCD22392218
  • Cas Number : 1421601-19-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112702

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112702
Availability details for
Quantity
  • Availability
  • Add to Cart