ENA372572596 Enamine

methyl 2-[4-(1-hydroxyethyl)-1H-1,2,3-triazol-1-yl]acetate

  • MDL number MFCD22392219
  • Cas Number : 1421603-55-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112724

Safety Information

msds https://www.enaminestore.com/catalog/EN300-112724
Availability details for
Quantity
  • Availability
  • Add to Cart