ENA372572625 Enamine

1-(4-ethoxypyrimidin-2-yl)-1,4-diazepane

  • MDL number MFCD22391947
  • Cas Number : 1394775-76-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106446

Safety Information

msds https://www.enaminestore.com/catalog/EN300-106446
Availability details for
Quantity
  • Availability
  • Add to Cart