ENA372572627 Enamine

1,4-diazabicyclo[2.2.2]octane-2-carbonitrile

  • MDL number MFCD22391914
  • Cas Number : 1394774-46-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103084

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-103084
Availability details for
Quantity
  • Availability
  • Add to Cart