ENA372572632 Enamine

1-bromo-3-(1-chloroethyl)benzene

  • MDL number MFCD11186422
  • Cas Number : 19935-76-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103418

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-103418
Availability details for
Quantity
  • Availability
  • Add to Cart