ENA372572654 Enamine

[5-(2-methylphenyl)-1,2,4-oxadiazol-3-yl]methanamine

  • MDL number MFCD07364466
  • Cas Number : 946745-07-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109365

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109365
Availability details for
Quantity
  • Availability
  • Add to Cart