ENA372572665 Enamine

2,4-diethyl 3-ethyl-5-methyl-1H-pyrrole-2,4-dicarboxylate

  • MDL number MFCD00030394
  • Cas Number : 66414-04-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108...110
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109547

Safety Information

msds https://www.enaminestore.com/catalog/EN300-109547
Availability details for
Quantity
  • Availability
  • Add to Cart