ENA372572687 Enamine

4-bromothiophene-3-sulfonyl chloride

  • MDL number MFCD04115406
  • Cas Number : 111283-90-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104391

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-104391
Availability details for
Quantity
  • Availability
  • Add to Cart