ENA372572691 Enamine

3-[(1-methyl-4-nitro-1H-imidazol-5-yl)oxy]aniline

  • MDL number MFCD12857020
  • Cas Number : 1183060-20-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110227

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110227
Availability details for
Quantity
  • Availability
  • Add to Cart