ENA372572695 Enamine

ethyl 5-amino-1-(6-methylpyridazin-3-yl)-1H-pyrazole-4-carboxylate

  • MDL number MFCD09882261
  • Cas Number : 1177350-37-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110256

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110256
Availability details for
Quantity
  • Availability
  • Add to Cart