ENA372572713 Enamine

1-(2,4-dimethylphenyl)-4-phenyl-1H-imidazole-2-thiol

  • MDL number MFCD01848551
  • Cas Number : 325994-32-9
  • Molecular Weight : 280.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-111104

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111104
Availability details for
Quantity
  • Availability
  • Add to Cart