ENA372572727 Enamine

benzyl 3-sulfamoylpropanoate

  • MDL number MFCD22392071
  • Cas Number : 1394738-31-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111200

Safety Information

msds https://www.enaminestore.com/catalog/EN300-111200
Availability details for
Quantity
  • Availability
  • Add to Cart