ENA372572740 Enamine

thiane-2-carboxylic acid

  • MDL number MFCD18815183
  • Cas Number : 53916-77-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107433

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107433
Availability details for
Quantity
  • Availability
  • Add to Cart