ENA372572744 Enamine

3-(4-bromophenyl)butanoic acid

  • MDL number MFCD08282290
  • Cas Number : 53086-46-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-111249

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111249
Availability details for
Quantity
  • Availability
  • Add to Cart