ENA372572747 Enamine

3-amino-4-[(2-methoxyethyl)amino]benzene-1-sulfonamide

  • MDL number MFCD11206640
  • Cas Number : 1094243-21-6
  • Molecular Weight : 245.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-111285

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111285
Availability details for
Quantity
  • Availability
  • Add to Cart