ENA372572748 Enamine

1-(propan-2-yl)-1H,4H,5H-pyrazolo[3,4-d]pyrimidin-4-one

  • MDL number MFCD14609282
  • Cas Number : 3763-25-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111286

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111286
Availability details for
Quantity
  • Availability
  • Add to Cart