ENA372572766 Enamine

2-fluoro-6-(1H-1,2,4-triazol-1-yl)pyridine

  • MDL number MFCD14600319
  • Cas Number : 1248407-47-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111332

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111332
Availability details for
Quantity
  • Availability
  • Add to Cart