ENA372572767 Enamine

5-phenyl-2,3-dihydro-1H-indole

  • MDL number MFCD06802121
  • Cas Number : 104636-54-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111333

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111333
Availability details for
Quantity
  • Availability
  • Add to Cart